Anti-AVPI1 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: AVPI1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77828-962
- 77828-963
- Antigen symbol:
- AVPI1
- AVPI1
- Antigen name:
- Arginine vasopressin-induced protein 1 protein (1-147AA)
- Arginine vasopressin-induced protein 1 protein (1-147AA)
- Antigen synonyms:
- NPD013|PP5395|Arginine vasopressin-induced protein 1|AVP-induced protein 1
- NPD013|PP5395|Arginine vasopressin-induced protein 1|AVP-induced protein 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5T686
- Q5T686
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together