Anti-COX17 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: COX17
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-941
- 77823-942
- Antigen symbol:
- COX17
- COX17
- Antigen name:
- Cytochrome c oxidase copper chaperone protein (1-63AA)
- Cytochrome c oxidase copper chaperone protein (1-63AA)
- Antigen synonyms:
- COX 17|COX17|COX17 cytochrome c oxidase assembly homolog (S. cerevisiae)|COX17 cytochrome c oxidase assembly homolog|COX17 homolog cytochrome c oxidase assembly protein|COX17_HUMAN|cytochrome c oxidase assembly protein cox17 homolog|Cytochrome c oxidase copper chaperone|Human homolog of yeast mitochondrial copper recruitment|MGC104397|MGC117386|OTTHUMP00000215284|OTTHUMP00000215285
- COX 17|COX17|COX17 cytochrome c oxidase assembly homolog (S. cerevisiae)|COX17 cytochrome c oxidase assembly homolog|COX17 homolog cytochrome c oxidase assembly protein|COX17_HUMAN|cytochrome c oxidase assembly protein cox17 homolog|Cytochrome c oxidase copper chaperone|Human homolog of yeast mitochondrial copper recruitment|MGC104397|MGC117386|OTTHUMP00000215284|OTTHUMP00000215285
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14061
- Q14061
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together