Anti-rbm24 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: rbm24
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Specifications
- Catalog No:
- 77829-865
- 77829-864
- Antigen symbol:
- rbm24
- rbm24
- Antigen name:
- Recombinant Zebrafish RNA-binding protein 24 protein (2-78AA)
- Recombinant Zebrafish RNA-binding protein 24 protein (2-78AA)
- Antigen synonyms:
- rbm24|RNA-binding protein 24|RNA-binding motif protein 24|RNA-binding protein SEB-4
- rbm24|RNA-binding protein 24|RNA-binding motif protein 24|RNA-binding protein SEB-4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Zebrafish
- Zebrafish
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q76LC6
- Q76LC6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together