Anti-IGFN1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77812-565
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Recombinant Protein
- Antigen symbol:IGFN1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q86VF2
- Antigen synonyms:IGFN1|EEF1A2BP1|KYIP1Immunoglobulin-like and fibronectin type III domain-containing protein 1|EEF1A2-binding protein 1|KY-interacting protein 1
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:2000 to 5000, IHC: 1:200 to 500
Type:
Antigen: IGFN1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat