Anti-ACOT11 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ACOT11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-703
- 77833-702
- Antigen symbol:
- ACOT11
- ACOT11
- Antigen name:
- Acyl-coenzyme A thioesterase 11 protein (35-254AA)
- Acyl-coenzyme A thioesterase 11 protein (35-254AA)
- Antigen synonyms:
- ACO11_HUMAN|ACOT 11|ACOT11|Acyl CoA thioester hydrolase 11|Acyl CoA thioesterase 11|Acyl coenzyme A thioesterase 11|Acyl-CoA thioester hydrolase 11|Acyl-CoA thioesterase 11|Acyl-coenzyme A thioesterase 11|Adipose associated thioesterase|Adipose-associated thioesterase|BFIT 1|BFIT 2|BFIT|BFIT1|BFIT2|Brown fat inducible thioesterase|Brown fat-inducible thioesterase|KIAA0707|OTTHUMP00000046722|StAR related lipid transfer (START) domain containing 14|STARD 14|STARD14|START domain containing 14|THEA|THEM 1|THEM1|Thioesterase adipose associated|Thioesterase superfamily member 1
- ACO11_HUMAN|ACOT 11|ACOT11|Acyl CoA thioester hydrolase 11|Acyl CoA thioesterase 11|Acyl coenzyme A thioesterase 11|Acyl-CoA thioester hydrolase 11|Acyl-CoA thioesterase 11|Acyl-coenzyme A thioesterase 11|Adipose associated thioesterase|Adipose-associated thioesterase|BFIT 1|BFIT 2|BFIT|BFIT1|BFIT2|Brown fat inducible thioesterase|Brown fat-inducible thioesterase|KIAA0707|OTTHUMP00000046722|StAR related lipid transfer (START) domain containing 14|STARD 14|STARD14|START domain containing 14|THEA|THEM 1|THEM1|Thioesterase adipose associated|Thioesterase superfamily member 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WXI4
- Q8WXI4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together