Anti-RANBP17 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: RANBP17
Clonality: Recombinant
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77838-334
- 77838-333
- Antigen symbol:
- RANBP17
- RANBP17
- Antigen name:
- Ran-binding protein 17 protein (465-587AA)
- Ran-binding protein 17 protein (465-587AA)
- Antigen synonyms:
- FLJ32916|RAN binding protein 17|Ran-binding protein 17|RanBP 17|Ranbp17|RBP17_HUMAN
- FLJ32916|RAN binding protein 17|Ran-binding protein 17|RanBP 17|Ranbp17|RBP17_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H2T7
- Q9H2T7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together