Anti-AOC3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: AOC3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77825-535
- 77825-536
- Antigen symbol:
- AOC3
- AOC3
- Antigen name:
- Membrane primary amine oxidase protein (27-259AA)
- Membrane primary amine oxidase protein (27-259AA)
- Antigen synonyms:
- Amine oxidase|copper containing 3 (vascular adhesion protein 1)|AOC3|AOC3_HUMAN|Copper amine oxidase|HPAO|Membrane copper amine oxidase|Membrane primary amine oxidase|Semicarbazide sensitive amine oxidase|Semicarbazide-sensitive amine oxidase|SSAO|VAP-1|Vascular adhesion protein 1|VP97
- Amine oxidase|copper containing 3 (vascular adhesion protein 1)|AOC3|AOC3_HUMAN|Copper amine oxidase|HPAO|Membrane copper amine oxidase|Membrane primary amine oxidase|Semicarbazide sensitive amine oxidase|Semicarbazide-sensitive amine oxidase|SSAO|VAP-1|Vascular adhesion protein 1|VP97
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16853
- Q16853
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together