Anti-PPP1R12C Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PPP1R12C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77838-170
- 77838-169
- Antigen symbol:
- PPP1R12C
- PPP1R12C
- Antigen name:
- Protein phosphatase 1 regulatory subunit 12C protein (317-450AA)
- Protein phosphatase 1 regulatory subunit 12C protein (317-450AA)
- Antigen synonyms:
- LENG 3|LENG3|leukocyte receptor cluster (LRC) encoded novel gene 3|leukocyte receptor cluster (LRC) member 3|MBS 85|MBS85|Myosin binding subunit 85|p84|p85|PP12C_HUMAN|Ppp1r12c|protein phosphatase 1 regulatory (inhibitor) subunit 12C|protein phosphatase 1 myosin binding subunit of 85 kDa|protein phosphatase 1 myosin binding subunit p85|Protein phosphatase 1 myosin-binding subunit of 85 kDa|Protein phosphatase 1 myosin-binding subunit p85|Protein phosphatase 1 regulatory subunit 12C
- LENG 3|LENG3|leukocyte receptor cluster (LRC) encoded novel gene 3|leukocyte receptor cluster (LRC) member 3|MBS 85|MBS85|Myosin binding subunit 85|p84|p85|PP12C_HUMAN|Ppp1r12c|protein phosphatase 1 regulatory (inhibitor) subunit 12C|protein phosphatase 1 myosin binding subunit of 85 kDa|protein phosphatase 1 myosin binding subunit p85|Protein phosphatase 1 myosin-binding subunit of 85 kDa|Protein phosphatase 1 myosin-binding subunit p85|Protein phosphatase 1 regulatory subunit 12C
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BZL4
- Q9BZL4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together