Anti-ZNF367 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: ZNF367
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77805-360
- 77805-361
- Antigen symbol:
- ZNF367
- ZNF367
- Antigen name:
- Zinc finger protein 367 protein (83-151AA)
- Zinc finger protein 367 protein (83-151AA)
- Antigen synonyms:
- C2H2 zinc finger protein ZFF29|Zinc finger protein 367|ZN367_HUMAN
- C2H2 zinc finger protein ZFF29|Zinc finger protein 367|ZN367_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7RTV3
- Q7RTV3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together