Anti-Nqo2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: Nqo2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Specifications
- Catalog No:
- 77828-332
- 77828-331
- Antigen symbol:
- Nqo2;Ribosyldihydronicotinamide dehydrogenase [quinone];EC 1.10.5.1;NRH dehydrogenase [quinone] 2;NRH:quinone oxidoreductase 2;Quinone reductase 2;QR2
- Nqo2;Ribosyldihydronicotinamide dehydrogenase [quinone];EC 1.10.5.1;NRH dehydrogenase [quinone] 2;NRH:quinone oxidoreductase 2;Quinone reductase 2;QR2
- Antigen name:
- Recombinant Rat Ribosyldihydronicotinamide dehydrogenase [quinone] protein (1-231AA)
- Recombinant Rat Ribosyldihydronicotinamide dehydrogenase [quinone] protein (1-231AA)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Rat
- Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6AY80
- Q6AY80
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together