Anti-FZD1 Rabbit Polyclonal Antibody
Catalog # 77809-339
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Synthesized peptide derived from the N-terminal region of Frizzled-1.
- Antigen symbol:FZD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9UP38
- Antigen synonyms:Chfz-1|Dfrizzled-1|DKFZp564G072|FLJ95923|Frizzled 1|Frizzled homolog 1 (Drosophila)|Frizzled homolog 1|Frizzled-1|Fz receptor 1|Fz-1|Fz1|FZD1|FZD1_HUMAN|FzE1|hFz1|Rfz1|Wnt receptor|Xfrizzled-1
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IF: 1:200 to 1:1000, ELISA: 1:20000
Type:
Antigen: FZD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human