Anti-PLA2G16 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PLA2G16
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77794-295
- 77794-296
- Antigen symbol:
- PLA2G16
- PLA2G16
- Antigen name:
- HRAS-like suppressor 3 protein (1-132AA)
- HRAS-like suppressor 3 protein (1-132AA)
- Antigen synonyms:
- Adipose specific phospholipase A2|Adipose specific PLA2|Adipose-specific phospholipase A2|AdPLA|Ca independent phospholipase A1/2|Group XVI phospholipase A1/A2|Group XVI phospholipase A2|H rev 107 protein homolog|H REV107 1|H-rev 107 protein homolog|HRAS like suppressor 1|HRAS like suppressor 3|HRAS-like suppressor 1|HRAS-like suppressor 3|HREV107 1|HREV107 3|HREV107|HREV107-1|HREV107-3|HRSL3|HRSL3_HUMAN|Phospholipase A2 group XVI|Renal carcinoma antigen NY REN 65|Renal carcinoma antigen NY-REN-65
- Adipose specific phospholipase A2|Adipose specific PLA2|Adipose-specific phospholipase A2|AdPLA|Ca independent phospholipase A1/2|Group XVI phospholipase A1/A2|Group XVI phospholipase A2|H rev 107 protein homolog|H REV107 1|H-rev 107 protein homolog|HRAS like suppressor 1|HRAS like suppressor 3|HRAS-like suppressor 1|HRAS-like suppressor 3|HREV107 1|HREV107 3|HREV107|HREV107-1|HREV107-3|HRSL3|HRSL3_HUMAN|Phospholipase A2 group XVI|Renal carcinoma antigen NY REN 65|Renal carcinoma antigen NY-REN-65
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P53816
- P53816
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together