Anti-KRT33B Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KRT33B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-410
- 77824-409
- Antigen symbol:
- KRT33B
- KRT33B
- Antigen name:
- Keratin, type I cuticular Ha3-II protein (1-404AA)
- Keratin, type I cuticular Ha3-II protein (1-404AA)
- Antigen synonyms:
- HA 3II|HA3II|Hair keratin|Hair keratin type I Ha3 II|Hard keratin type I 3II|hHa3 II|hHa3II|HKA3B|K33B|Keratin 33B|Keratin|Keratin hair acidic 3B|Keratin hair acidic3B|Keratin type I cuticular Ha3 II|Keratin-33B|Keratin33B|KRT 33B|KRT33B|KRTHA 3B|KRTHA3A|KRTHA3B|KT33B_HUMAN|type I cuticular Ha3-II|type I Ha3-II|Type I hair keratin 3B
- HA 3II|HA3II|Hair keratin|Hair keratin type I Ha3 II|Hard keratin type I 3II|hHa3 II|hHa3II|HKA3B|K33B|Keratin 33B|Keratin|Keratin hair acidic 3B|Keratin hair acidic3B|Keratin type I cuticular Ha3 II|Keratin-33B|Keratin33B|KRT 33B|KRT33B|KRTHA 3B|KRTHA3A|KRTHA3B|KT33B_HUMAN|type I cuticular Ha3-II|type I Ha3-II|Type I hair keratin 3B
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14525
- Q14525
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together