Anti-BABAM2 Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: BABAM2
Clonality: Recombinant
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77838-852
- 77838-851
- Antigen symbol:
- BABAM2
- BABAM2
- Antigen name:
- BRISC and BRCA1-A complex member 2 protein (94-383AA)
- BRISC and BRCA1-A complex member 2 protein (94-383AA)
- Antigen synonyms:
- 6030405P19Rik|AI429776|B830038C02Rik|Brain and reproductive organ expressed (TNFRSF1A modulator)|brain and reproductive organ expressed protein|Brain and reproductive organ-expressed protein|BRCA1 A complex subunit BRE|BRCA1-A complex subunit BRE|BRCA1/BRCA2 containing complex subunit 4|BRCA1/BRCA2 containing complex subunit 45|BRCA1/BRCA2-containing complex subunit 45|BRCC4|BRCC45|bre|BRE_HUMAN
- 6030405P19Rik|AI429776|B830038C02Rik|Brain and reproductive organ expressed (TNFRSF1A modulator)|brain and reproductive organ expressed protein|Brain and reproductive organ-expressed protein|BRCA1 A complex subunit BRE|BRCA1-A complex subunit BRE|BRCA1/BRCA2 containing complex subunit 4|BRCA1/BRCA2 containing complex subunit 45|BRCA1/BRCA2-containing complex subunit 45|BRCC4|BRCC45|bre|BRE_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NXR7
- Q9NXR7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together