Anti-SH3GLB1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SH3GLB1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77839-691
- 77839-692
- Antigen symbol:
- SH3GLB1
- SH3GLB1
- Antigen name:
- Endophilin-B1 protein (1-365AA)
- Endophilin-B1 protein (1-365AA)
- Antigen synonyms:
- Bax interacting factor 1|Bax-interacting factor 1|Bif 1|Bif|Bif-1|CGI 61|dJ612B15.2|Endophilin B1|Endophilin-B1|PPP1R70|Protein phosphatase 1 regulatory subunit 70|SH3 containing protein SH3GLB1|SH3 domain GRB2 like endophilin B1|SH3 domain-containing GRB2-like protein B1|Sh3glb1|SH3p13|SHLB1_HUMAN
- Bax interacting factor 1|Bax-interacting factor 1|Bif 1|Bif|Bif-1|CGI 61|dJ612B15.2|Endophilin B1|Endophilin-B1|PPP1R70|Protein phosphatase 1 regulatory subunit 70|SH3 containing protein SH3GLB1|SH3 domain GRB2 like endophilin B1|SH3 domain-containing GRB2-like protein B1|Sh3glb1|SH3p13|SHLB1_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y371
- Q9Y371
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together