Anti-entD Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: entD
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Staphylococcus aureus
- Catalog No:
- 77815-547
- 77815-546
- Antigen symbol:
- entD
- entD
- Antigen name:
- Recombinant Staphylococcus aureus Enterotoxin type D protein (26-258AA)
- Recombinant Staphylococcus aureus Enterotoxin type D protein (26-258AA)
- Antigen synonyms:
- entDEnterotoxin type D|SED
- entDEnterotoxin type D|SED
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Staphylococcus aureus
- Staphylococcus aureus
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P20723
- P20723
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together