Anti-ZNF165 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: ZNF165
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77805-034
- 77805-033
- Antigen symbol:
- ZNF165
- ZNF165
- Antigen name:
- Zinc finger protein 165 protein (1-485AA)
- Zinc finger protein 165 protein (1-485AA)
- Antigen synonyms:
- Cancer/testis antigen 53|CT53|LD65|MGC140342|OTTHUMP00000016195|Zinc finger and SCAN domain containing protein 7|Zinc finger and SCAN domain-containing protein 7|Zinc finger protein 165|ZINC FINGER PROTEIN 165, ZNF165|ZN165_HUMAN|ZNF165|ZPF165|ZSCAN7
- Cancer/testis antigen 53|CT53|LD65|MGC140342|OTTHUMP00000016195|Zinc finger and SCAN domain containing protein 7|Zinc finger and SCAN domain-containing protein 7|Zinc finger protein 165|ZINC FINGER PROTEIN 165, ZNF165|ZN165_HUMAN|ZNF165|ZPF165|ZSCAN7
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P49910
- P49910
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together