Anti-SLC13A5 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC13A5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77831-779
- 77831-778
- Antigen symbol:
- SLC13A5
- SLC13A5
- Antigen name:
- Solute carrier family 13 member 5 protein (156-204AA)
- Solute carrier family 13 member 5 protein (156-204AA)
- Antigen synonyms:
- Na(+)/citrate cotransporter| NaC2/NaCT| NaCT| Novel solute carrier family 13 (sodium dependent dicarboxylate transporter) (Slc13a2 or 3) member| S13A5_HUMAN| Slc13a5| Sodium coupled citrate transporter| Sodium-coupled citrate transporter| Sodium-dependent citrate transporter| Solute carrier family 13 (sodium dependent citrate transporter)| member 5| Solute carrier family 13 member 5
- Na(+)/citrate cotransporter| NaC2/NaCT| NaCT| Novel solute carrier family 13 (sodium dependent dicarboxylate transporter) (Slc13a2 or 3) member| S13A5_HUMAN| Slc13a5| Sodium coupled citrate transporter| Sodium-coupled citrate transporter| Sodium-dependent citrate transporter| Solute carrier family 13 (sodium dependent citrate transporter)| member 5| Solute carrier family 13 member 5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86YT5
- Q86YT5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together