Anti-Genome polyprotein Rabbit Recombinant Antibody
New Product
Type:
Antigen: Genome polyprotein
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human rhinovirus A serotype 89
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77817-532
- 77817-533
- Antigen symbol:
- Genome polyprotein
- Genome polyprotein
- Antigen name:
- rhinovirus A serotype 89 Genome polyprotein protein (575-866AA)
- rhinovirus A serotype 89 Genome polyprotein protein (575-866AA)
- Antigen synonyms:
- Genome polyprotein [Cleaved into: P1|Capsid protein VP0|VP4-VP2)|Capsid protein VP4|P1A|Virion protein 4)|Capsid protein VP2|P1B|Virion protein 2)|Capsid protein VP3|P1C|Virion protein 3)|Capsid protein VP1|P1D|Virion protein 1)|P2|Protease 2A|P2A|EC 3.4.22.29|Picornain 2A|Protein 2A)|Protein 2B|P2B)|Protein 2C|P2C|EC 3.6.1.15)|P3|Protein 3AB|Protein 3A|P3A)|Viral protein genome-linked|VPg|Protein 3B|P3B)|Protein 3CD|EC 3.4.22.28)|Protease 3C|EC 3.4.22.28|Picornain 3C|P3C)|RNA-directed RNA polymerase|RdRp|EC 2.7.7.48|3D polymerase|3Dpol|Protein 3D|3D)]
- Genome polyprotein [Cleaved into: P1|Capsid protein VP0|VP4-VP2)|Capsid protein VP4|P1A|Virion protein 4)|Capsid protein VP2|P1B|Virion protein 2)|Capsid protein VP3|P1C|Virion protein 3)|Capsid protein VP1|P1D|Virion protein 1)|P2|Protease 2A|P2A|EC 3.4.22.29|Picornain 2A|Protein 2A)|Protein 2B|P2B)|Protein 2C|P2C|EC 3.6.1.15)|P3|Protein 3AB|Protein 3A|P3A)|Viral protein genome-linked|VPg|Protein 3B|P3B)|Protein 3CD|EC 3.4.22.28)|Protease 3C|EC 3.4.22.28|Picornain 3C|P3C)|RNA-directed RNA polymerase|RdRp|EC 2.7.7.48|3D polymerase|3Dpol|Protein 3D|3D)]
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human rhinovirus A serotype 89
- Human rhinovirus A serotype 89
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P07210
- P07210
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together