Anti-RGS20 Rabbit Polyclonal Antibody
Catalog # 77797-214
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:ZGAP1
- Antigen symbol:RGS20
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O76081
- Antigen synonyms:G(z)GAP|Gz selective GTPase activating protein|Gz-GAP|Gz-selective GTPase-activating protein|HGNC:14600|Regulator of G protein signaling 20|Regulator of G-protein signaling 20|Regulator of G-protein signaling Z1|Regulator of Gz selective protein signaling 1|Regulator of Gz-selective protein signaling 1|RGS20|RGS20_HUMAN|RGSZ1|ZGAP1
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: RGS20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat