Anti-TSPAN15 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: TSPAN15
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-628
- 77803-628
- Antigen symbol:
- TSPAN15
- TSPAN15
- Antigen name:
- Tetraspanin-15 protein (115-235AA)
- Tetraspanin-15 protein (115-235AA)
- Antigen synonyms:
- Tetraspan NET-7|Tetraspanin 15|Tetraspanin-15|Transmembrane 4 superfamily member 15|TSN15_HUMAN|Tspan-15|TSPAN15
- Tetraspan NET-7|Tetraspanin 15|Tetraspanin-15|Transmembrane 4 superfamily member 15|TSN15_HUMAN|Tspan-15|TSPAN15
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95858
- O95858
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together