Anti-GPR174 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: GPR174
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77842-939
- 77842-938
- Antigen symbol:
- GPR174
- GPR174
- Antigen name:
- Probable G-protein coupled receptor 174 protein (291-333AA)
- Probable G-protein coupled receptor 174 protein (291-333AA)
- Antigen synonyms:
- FKSG79|G protein-coupled receptor 174|GP174_HUMAN|Gpr174|GRPC 174|GRPC174|jeg18|Probable G-protein coupled receptor 174|purinergic receptor fksg79|Putative purinergic receptor|Putative purinergic receptor FKSG792
- FKSG79|G protein-coupled receptor 174|GP174_HUMAN|Gpr174|GRPC 174|GRPC174|jeg18|Probable G-protein coupled receptor 174|purinergic receptor fksg79|Putative purinergic receptor|Putative purinergic receptor FKSG792
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BXC1
- Q9BXC1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together