Order Entry
Canada
ContactUsLinkComponent
Anti-CENPX Rabbit Recombinant Polyclonal Antibody
Anti-CENPX Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-CENPX Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 77801-400
  • 77801-399
  • Antigen symbol:
  • CENPX
  • CENPX
  • Antigen name:
  • Centromere protein X protein (1-63AA)
  • Centromere protein X protein (1-63AA)
  • Antigen synonyms:
  • CENP X|CENP-X|CENPX|CENPX_HUMAN|Centromere protein X|FAAP10|FANCM interacting histone fold protein 2|FANCM-interacting histone fold protein 2|Fanconi anemia associated polypeptide of 10 kDa|Fanconi anemia-associated polypeptide of 10 kDa|MGC14480|Retinoic acid inducible gene D9 protein homolog|Retinoic acid-inducible gene D9 protein homolog|Stimulated by retinoic acid gene 13 protein homolog|STRA13
  • CENP X|CENP-X|CENPX|CENPX_HUMAN|Centromere protein X|FAAP10|FANCM interacting histone fold protein 2|FANCM-interacting histone fold protein 2|Fanconi anemia associated polypeptide of 10 kDa|Fanconi anemia-associated polypeptide of 10 kDa|MGC14480|Retinoic acid inducible gene D9 protein homolog|Retinoic acid-inducible gene D9 protein homolog|Stimulated by retinoic acid gene 13 protein homolog|STRA13
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • A8MT69
  • A8MT69
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together