Anti-DEFB126 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: DEFB126
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77836-933
- 77836-932
- Antigen symbol:
- DEFB126
- DEFB126
- Antigen name:
- Beta-defensin 126 protein (36-111AA)
- Beta-defensin 126 protein (36-111AA)
- Antigen synonyms:
- DEFB126|C20orf8|DEFB26Beta-defensin 126|Beta-defensin 26|DEFB-26|Defensin|beta 126|Epididymal secretory protein 13.2|ESP13.2|HBD26
- DEFB126|C20orf8|DEFB26Beta-defensin 126|Beta-defensin 26|DEFB-26|Defensin|beta 126|Epididymal secretory protein 13.2|ESP13.2|HBD26
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BYW3
- Q9BYW3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together