Anti-S100A9 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: S100A9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77798-552
- 77798-553
- Antigen symbol:
- S100A9
- S100A9
- Antigen name:
- Protein S100-A9 protein (2-114AA)
- Protein S100-A9 protein (2-114AA)
- Antigen synonyms:
- Leukocyte L1 complex heavy chain|60B8AG|CAGB|Calgranulin B|Calgranulin-B|Calprotectin L1H subunit|CFAG|CGLB|Cystic fibrosis antigen B|L1AG|LIAG|MAC387|MIF|Migration inhibitory factor related protein 14|Migration inhibitory factor-related protein 14|MRP 14|MRP-14|MRP14|Myeloid-related protein 14|NIF|OTTHUMP00000015331|p14|Protein S100-A9|S100 A9|S100 calcium binding protein A9|S100 calcium binding protein A9 calgranulin B|S100 calcium-binding protein A9|S100A9|S10A9_HUMAN
- Leukocyte L1 complex heavy chain|60B8AG|CAGB|Calgranulin B|Calgranulin-B|Calprotectin L1H subunit|CFAG|CGLB|Cystic fibrosis antigen B|L1AG|LIAG|MAC387|MIF|Migration inhibitory factor related protein 14|Migration inhibitory factor-related protein 14|MRP 14|MRP-14|MRP14|Myeloid-related protein 14|NIF|OTTHUMP00000015331|p14|Protein S100-A9|S100 A9|S100 calcium binding protein A9|S100 calcium binding protein A9 calgranulin B|S100 calcium-binding protein A9|S100A9|S10A9_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P06702
- P06702
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together