Anti-TAF6 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TAF6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77801-813
- 77801-812
- Antigen symbol:
- TAF6
- TAF6
- Antigen name:
- Transcription initiation factor TFIID subunit 6 protein (627-677AA)
- Transcription initiation factor TFIID subunit 6 protein (627-677AA)
- Antigen synonyms:
- TAF2E|TAFII70|Transcription initiation factor TFIID subunit 6|RNA polymerase II TBP-associated factor subunit E|Transcription initiation factor TFIID 70 kDa subunit|TAF(II)70|TAFII-70|Transcription initiation factor TFIID 80 kDa subunit|TAF(II)80|TAFII-80|TAFII80
- TAF2E|TAFII70|Transcription initiation factor TFIID subunit 6|RNA polymerase II TBP-associated factor subunit E|Transcription initiation factor TFIID 70 kDa subunit|TAF(II)70|TAFII-70|Transcription initiation factor TFIID 80 kDa subunit|TAF(II)80|TAFII-80|TAFII80
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P49848
- P49848
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together