Anti-TRIM37 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TRIM37
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-547
- 77803-547
- Antigen symbol:
- TRIM37
- TRIM37
- Antigen name:
- E3 ubiquitin-protein ligase TRIM37 protein (301-600AA)
- E3 ubiquitin-protein ligase TRIM37 protein (301-600AA)
- Antigen synonyms:
- E3 ubiquitin protein ligase TRIM37|E3 ubiquitin-protein ligase TRIM37|KIAA0898|MUL|MUL protein|Mulibrey nanism gene|Mulibrey nanism protein|POB 1|POB1|RING B box coiled coil protein|TEF 3|TEF3|TRI37_HUMAN|TRIM 37|Trim37|Tripartite motif containing 37|Tripartite motif containing 37 protein|Tripartite motif containing protein 37|Tripartite motif-containing protein 37
- E3 ubiquitin protein ligase TRIM37|E3 ubiquitin-protein ligase TRIM37|KIAA0898|MUL|MUL protein|Mulibrey nanism gene|Mulibrey nanism protein|POB 1|POB1|RING B box coiled coil protein|TEF 3|TEF3|TRI37_HUMAN|TRIM 37|Trim37|Tripartite motif containing 37|Tripartite motif containing 37 protein|Tripartite motif containing protein 37|Tripartite motif-containing protein 37
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O94972
- O94972
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together