Anti-NPC2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NPC2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog
Specifications
- Catalog No:
- 77825-628
- 77825-629
- Antigen symbol:
- NPC2
- NPC2
- Antigen name:
- Recombinant Dog NPC intracellular cholesterol transporter 2 protein (22-149AA)
- Recombinant Dog NPC intracellular cholesterol transporter 2 protein (22-149AA)
- Antigen synonyms:
- NPC2|NPC intracellular cholesterol transporter 2|Epididymal secretory protein E1|cE1|Niemann Pick type C2 protein homolog
- NPC2|NPC intracellular cholesterol transporter 2|Epididymal secretory protein E1|cE1|Niemann Pick type C2 protein homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Dog
- Dog
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q28895
- Q28895
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together