Anti-ECI2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: ECI2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat;Mouse
Specifications
- Catalog No:
- 77819-457
- 77819-458
- Antigen symbol:
- ECI2
- ECI2
- Antigen name:
- Enoyl-CoA delta isomerase 2, mitochondrial protein (119-219AA)
- Enoyl-CoA delta isomerase 2, mitochondrial protein (119-219AA)
- Antigen synonyms:
- 2-trans-enoyl-CoA isomerase|ACBD2|acyl Coenzyme A binding domain containing 2|D2-enoyl-CoA isomerase|D3|D3 D2 enoyl CoA isomerase|DBI related protein 1|DBI-related protein 1|delta(2)-enoyl-CoA isomerase|Delta(3)|Delta(3) delta(2) enoyl CoA isomerase|Diazepam-binding inhibitor-related protein 1|Dodecenoyl CoA isomerase|Dodecenoyl-CoA isomerase|DRS 1|DRS-1|DRS1|Eci2|ECI2_HUMAN|Enoyl-CoA delta isomerase 2|HCA88|Hepatocellular carcinoma associated antigen 88|Hepatocellular carcinoma-associated antigen 88|mitochondrial|pECI|Peroxisomal 3|Peroxisomal 3,2-trans-enoyl-CoA isomerase|Peroxisomal D3 D2 enoyl CoA isomerase|Renal carcinoma antigen NY REN 1|Renal carcinoma antigen NY-REN-1
- 2-trans-enoyl-CoA isomerase|ACBD2|acyl Coenzyme A binding domain containing 2|D2-enoyl-CoA isomerase|D3|D3 D2 enoyl CoA isomerase|DBI related protein 1|DBI-related protein 1|delta(2)-enoyl-CoA isomerase|Delta(3)|Delta(3) delta(2) enoyl CoA isomerase|Diazepam-binding inhibitor-related protein 1|Dodecenoyl CoA isomerase|Dodecenoyl-CoA isomerase|DRS 1|DRS-1|DRS1|Eci2|ECI2_HUMAN|Enoyl-CoA delta isomerase 2|HCA88|Hepatocellular carcinoma associated antigen 88|Hepatocellular carcinoma-associated antigen 88|mitochondrial|pECI|Peroxisomal 3|Peroxisomal 3,2-trans-enoyl-CoA isomerase|Peroxisomal D3 D2 enoyl CoA isomerase|Renal carcinoma antigen NY REN 1|Renal carcinoma antigen NY-REN-1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75521
- O75521
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together