Anti-USP38 Rabbit Polyclonal Antibody
Catalog # 77804-144
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:KIAA1891
- Antigen symbol:USP38
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q8NB14
- Antigen synonyms:KIAA1891|Deubiquitinating enzyme 38|HP43.8KD|Ubiquitin carboxyl terminal hydrolase 38|Ubiquitin carboxyl-terminal hydrolase 38|Ubiquitin specific peptidase 38|Ubiquitin specific processing protease 38|Ubiquitin thioesterase 38|Ubiquitin thiolesterase 38|Ubiquitin-specific-processing protease 38|UBP38_HUMAN|USP 38|USP38
- Storage buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: USP38
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat