Anti-SIGIRR Rabbit Recombinant Polyclonal Antibody
Catalog # 77829-630
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Single Ig IL-1-related receptor protein (1-120AA)
- Antigen symbol:SIGIRR
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q6IA17
- Antigen synonyms:Ig IL1 related receptor| MGC110992| SIGIR_HUMAN| SIGIRR| Single Ig IL 1 related receptor| Single Ig IL-1-related receptor| Single Ig IL-1R-related molecule| Single Ig IL1R Related Molecule| Single immunoglobulin and toll interleukin 1 receptor (TIR) domain| Single immunoglobulin and toll interleukin 1 receptor domain| Single immunoglobulin domain containing IL1R related protein| Single immunoglobulin domain IL1R1 related| Single immunoglobulin domain-containing IL1R-related protein| TIR 8| TIR8| Toll/interleukin 1 receptor 8| Toll/interleukin-1 receptor 8
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: SIGIRR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse