Anti-SLC2A4 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC2A4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-988
- 77799-989
- Antigen symbol:
- SLC2A4
- SLC2A4
- Antigen name:
- Solute carrier family 2, facilitated glucose transporter member 4 protein (223-287AA)
- Solute carrier family 2, facilitated glucose transporter member 4 protein (223-287AA)
- Antigen synonyms:
- insulin-responsive|Glucose transporter GLUT 4|Glucose transporter type 4|Glucose transporter type 4 insulin responsive|GLUT 4|GLUT-4|GLUT4|GTR4_HUMAN|Insulin responsive glucose transporter type 4|kug|SLC 2A4|SLC2A4|solute carrier family 2 (facilitated glucose transporter) member 4|Solute carrier family 2 member 4|Solute carrier family 2, facilitated glucose transporter member 4
- insulin-responsive|Glucose transporter GLUT 4|Glucose transporter type 4|Glucose transporter type 4 insulin responsive|GLUT 4|GLUT-4|GLUT4|GTR4_HUMAN|Insulin responsive glucose transporter type 4|kug|SLC 2A4|SLC2A4|solute carrier family 2 (facilitated glucose transporter) member 4|Solute carrier family 2 member 4|Solute carrier family 2, facilitated glucose transporter member 4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P14672
- P14672
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together