Anti-ERBB3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: ERBB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77820-825
- 77820-824
- Antigen symbol:
- ERBB3
- ERBB3
- Antigen name:
- Fusion protein of ERBB3
- Fusion protein of ERBB3
- Antigen synonyms:
- c erbB 3|c erbB3|Erb b2 receptor tyrosine kinase 3|ErbB 3|ERBB3|ERBB3 protein|erbB3 S|ERBB3_HUMAN|Glial growth factor receptor|HER 3|HER3|Human epidermal growth factor receptor 3|LCCS2|MDA BF 1|MGC88033|p180 ErbB3|p45 sErbB3|p85 sErbB3|proto-oncogene-like protein c ErbB 3|proto-oncogene-like protein c ErbB3|Proto-oncogene-like protein c-ErbB-3|Receptor tyrosine protein kinase erbB 3|Receptor tyrosine protein kinase erbB3|Receptor tyrosine-protein kinase erbB-3|Tyrosine kinase type cell surface receptor HER3|Tyrosine kinase-type cell surface receptor HER3|v erb b2 avian erythroblastic leukemia viral oncogene homolog 3|v erb b2 erythroblastic leukemia viral oncogene homolog 3 (avian)|v erb b2 erythroblastic leukemia viral oncogene homolog 3
- c erbB 3|c erbB3|Erb b2 receptor tyrosine kinase 3|ErbB 3|ERBB3|ERBB3 protein|erbB3 S|ERBB3_HUMAN|Glial growth factor receptor|HER 3|HER3|Human epidermal growth factor receptor 3|LCCS2|MDA BF 1|MGC88033|p180 ErbB3|p45 sErbB3|p85 sErbB3|proto-oncogene-like protein c ErbB 3|proto-oncogene-like protein c ErbB3|Proto-oncogene-like protein c-ErbB-3|Receptor tyrosine protein kinase erbB 3|Receptor tyrosine protein kinase erbB3|Receptor tyrosine-protein kinase erbB-3|Tyrosine kinase type cell surface receptor HER3|Tyrosine kinase-type cell surface receptor HER3|v erb b2 avian erythroblastic leukemia viral oncogene homolog 3|v erb b2 erythroblastic leukemia viral oncogene homolog 3 (avian)|v erb b2 erythroblastic leukemia viral oncogene homolog 3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P21860
- P21860
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together