Anti-USP47 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: USP47
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
- Catalog No:
- 77807-787
- 77807-788
- Antigen symbol:
- USP47
- USP47
- Antigen name:
- Fusion protein of USP47
- Fusion protein of USP47
- Antigen synonyms:
- Deubiquitinating enzyme 47|TFRP|Trf TATA binding protein related factor proximal homolog|Trf TATA binding protein related factor proximal homolog Drosophila|Ubiquitin carboxyl-terminal hydrolase 47|Ubiquitin specific peptidase 47|Ubiquitin specific processing protease 47|Ubiquitin specific protease 47|Ubiquitin thioesterase 47|Ubiquitin thiolesterase 47|Ubiquitin-specific-processing protease 47|UBP47_HUMAN
- Deubiquitinating enzyme 47|TFRP|Trf TATA binding protein related factor proximal homolog|Trf TATA binding protein related factor proximal homolog Drosophila|Ubiquitin carboxyl-terminal hydrolase 47|Ubiquitin specific peptidase 47|Ubiquitin specific processing protease 47|Ubiquitin specific protease 47|Ubiquitin thioesterase 47|Ubiquitin thiolesterase 47|Ubiquitin-specific-processing protease 47|UBP47_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96K76
- Q96K76
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together