Anti-UIMC1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UIMC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-339
- 77834-338
- Antigen symbol:
- UIMC1
- UIMC1
- Antigen name:
- BRCA1-A complex subunit RAP80 protein (161-235AA)
- BRCA1-A complex subunit RAP80 protein (161-235AA)
- Antigen synonyms:
- BRCA1-A complex subunit RAP80|Nuclear zinc finger protein RAP80|OTTHUMP00000161441|OTTHUMP00000223372|OTTHUMP00000223374|RAP80|Receptor associated protein 80|Receptor-associated protein 80|Retinoid X receptor interacting protein 110|Retinoid x receptor interacting protein|Retinoid X receptor-interacting protein 110|RIP110|Rxrip110|Ubiquitin interaction motif containing 1|Ubiquitin interaction motif containing protein 1|Ubiquitin interaction motif-containing protein 1|UIMC1|UIMC1_HUMAN|X2HRIP110
- BRCA1-A complex subunit RAP80|Nuclear zinc finger protein RAP80|OTTHUMP00000161441|OTTHUMP00000223372|OTTHUMP00000223374|RAP80|Receptor associated protein 80|Receptor-associated protein 80|Retinoid X receptor interacting protein 110|Retinoid x receptor interacting protein|Retinoid X receptor-interacting protein 110|RIP110|Rxrip110|Ubiquitin interaction motif containing 1|Ubiquitin interaction motif containing protein 1|Ubiquitin interaction motif-containing protein 1|UIMC1|UIMC1_HUMAN|X2HRIP110
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96RL1
- Q96RL1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together