Anti-HSD11B1L Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: HSD11B1L
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77832-091
- 77832-092
- Antigen symbol:
- HSD11B1L
- HSD11B1L
- Antigen name:
- Hydroxysteroid 11-beta-dehydrogenase 1-like protein (17-87AA)
- Hydroxysteroid 11-beta-dehydrogenase 1-like protein (17-87AA)
- Antigen synonyms:
- HSD11B1L|HSD3|SCDR10|SDR26C2Hydroxysteroid 11-beta-dehydrogenase 1-like protein|EC 1.1.1.-|11-beta-hydroxysteroid dehydrogenase type 3|11-DH3|11-beta-HSD3|Short chain dehydrogenase/reductase family 26C member 2|Short-chain dehydrogenase/reductase 10
- HSD11B1L|HSD3|SCDR10|SDR26C2Hydroxysteroid 11-beta-dehydrogenase 1-like protein|EC 1.1.1.-|11-beta-hydroxysteroid dehydrogenase type 3|11-DH3|11-beta-HSD3|Short chain dehydrogenase/reductase family 26C member 2|Short-chain dehydrogenase/reductase 10
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7Z5J1
- Q7Z5J1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together