Anti-CNPY2 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CNPY2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77808-343
- 77808-342
- Antigen symbol:
- CNPY2
- CNPY2
- Antigen name:
- Protein canopy homolog 2 protein (22-182AA)
- Protein canopy homolog 2 protein (22-182AA)
- Antigen synonyms:
- Canopy 2 homolog (zebrafish)|Cnpy2|CNPY2_HUMAN|MIR-interacting saposin-like protein|MSAP|Protein canopy homolog 2|Putative secreted protein ZSIG9|TMEM4|Transmembrane protein 4|UNQ1943/PRO4426|ZSIG9
- Canopy 2 homolog (zebrafish)|Cnpy2|CNPY2_HUMAN|MIR-interacting saposin-like protein|MSAP|Protein canopy homolog 2|Putative secreted protein ZSIG9|TMEM4|Transmembrane protein 4|UNQ1943/PRO4426|ZSIG9
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y2B0
- Q9Y2B0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together