Anti-URI1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: URI1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77819-549
- 77819-550
- Antigen symbol:
- URI1
- URI1
- Antigen name:
- Unconventional prefoldin RPB5 interactor 1 protein (180-298AA)
- Unconventional prefoldin RPB5 interactor 1 protein (180-298AA)
- Antigen synonyms:
- C19orf2|Chromosome 19 open reading frame 2|NNX3|NNX3 protein|PPP1R19|Protein NNX3|Protein phosphatase 1 regulatory subunit 19|RMP|RMP_HUMAN|RNA polymerase II subunit 5 mediating protein|RNA polymerase II subunit 5-mediating protein|RPB5 mediating protein|RPB5-mediating protein|Unconventional prefoldin RPB5 interactor 1|Unconventional prefoldin RPB5 interactor|URI|URI1|URI1, prefoldin-like chaperone
- C19orf2|Chromosome 19 open reading frame 2|NNX3|NNX3 protein|PPP1R19|Protein NNX3|Protein phosphatase 1 regulatory subunit 19|RMP|RMP_HUMAN|RNA polymerase II subunit 5 mediating protein|RNA polymerase II subunit 5-mediating protein|RPB5 mediating protein|RPB5-mediating protein|Unconventional prefoldin RPB5 interactor 1|Unconventional prefoldin RPB5 interactor|URI|URI1|URI1, prefoldin-like chaperone
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O94763
- O94763
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together