CDN-D-2356
- Catalog No:
- 77860-528
- 77863-491
- CAS No.:
- 93951-79-2
- 93951-79-2
- Formula:
- C₆HD₄NO₃
- C₆HD₄NO₃
Product Family Options
- DescriptionSize
- : CDN-D-2356-0.1G
Specifications
Cat. No.77860-528CAS No.93951-79-2FormulaC₆HD₄NO₃ - : CDN-D-2356-0.25G
Specifications
Cat. No.77863-491CAS No.93951-79-2FormulaC₆HD₄NO₃
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



