Order Entry
Canada
Orders LinkContactUsLinkComponent
Hexafluorobenzene ≥99.0%

Hexafluorobenzene ≥99.0%

5 g
Catalog # TCH0085-5G
Supplier:   TCI America
Supplier #:  H0085-5G
Hexafluorobenzene ≥99.0%
Catalog # TCH0085-5G
Supplier:  TCI America
CAS Number:  392-56-3
Hexafluorobenzene ≥99.0%
Hexafluorobenzene ≥99.0%

5 g

CAS Number:  392-56-3
Price
$42.10
Log in for availability
Each (5g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

CAS Number: 392-56-3
MDL Number: MFCD00000288
Molecular Formula: C6F6
Molecular Weight: 186.06
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 81
Melting point (°C): 5
Flash Point (°C): 10
Specific Gravity (20/20): 1.62

Product Categories: Synthetic Organic Chemistry, Fluorous Chemistry, Fluorinated Building Blocks, Analytical Science, Chemical Shift Standards for Multinuclear NMR
Other Product Names / Synonyms: Perfluorobenzene

Synonyms:  Perfluorobenzene
  • CAS Number:392-56-3
  • Formula:C₆F₆
  • MDL Number:MFCD00000288
  • Molecular Weight:186.06 g/mol
  • Merck Index:13,04705
  • Boiling Pt:81 °C
  • Melting Pt:5 °C
  • Density:1.62 (20 °C)
  • Flash Pt:10 °C
  • Storage Temperature:Ambient
  • EINECS:206-876-2
  • UN:1993
  • ADR:3,II

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Purity/Analysis Method:
    99.0% (GC)

Specifications

  • Size:
    5 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.