Order Entry
Canada
Orders LinkContactUsLinkComponent
4,4'-Dimethylbenzil ≥99.0%

4,4'-Dimethylbenzil ≥99.0%

25 g
Catalog # TCD1875-025G
Supplier:   TCI America
Supplier #:  D1875-25G
4,4'-Dimethylbenzil ≥99.0%
Catalog # TCD1875-025G
Supplier:  TCI America
CAS Number:  3457-48-5
4,4'-Dimethylbenzil ≥99.0%
4,4'-Dimethylbenzil ≥99.0%

25 g

CAS Number:  3457-48-5
Price
$249.00
Log in for availability
Each (25g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS Number: 3457-48-5
MDL Number: MFCD00008554
Molecular Formula: C16H14O2
Molecular Weight: 238.29
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 211
Melting point (°C): 105

Synonyms:  1,2-Di-(4-methylphenyl)-1,2-ethanedione, Bis(4-methylphenyl)ethanedione, Di-p-tolylethanedione, 1,2-Di-p-tolylethane-1,2-dione
  • CAS Number:3457-48-5
  • Formula:C₁₆H₁₄O₂
  • MDL Number:MFCD00008554
  • Molecular Weight:238.29 g/mol
  • Boiling Pt:211 °C
  • Melting Pt:105 °C
  • EINECS:222-388-2

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Purity/Analysis Method:
    99.0% (GC)

Specifications

  • Size:
    25 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.