ContactUsLinkComponent
3B Scientific® BONElike™ Childs Vertebral Column
3B Scientific® BONElike™ Childs Vertebral Column
Catalog # 470137-712
undefined
3B Scientific® BONElike™ Childs Vertebral Column
Catalog # 470137-712
Supplier Number:  1000118
Specifications
  • Description:
    Childs Vertebral Column
  • Environmentally Preferable:
  • Cat. no.:
    470137-712
  • Supplier No.:
    A52
Specifications
About this item
This true-to-life anatomical replica of the vertebral column of a child of about 5 years old is especially interesting for those working in the areas of anatomy, pediatrics, orthopedics and pediatric radiology.
  • Compact display of the core skeletal structure
  • 3 year manufacturers warranty
  • Includes 3B smart anatomy
The unique material of the spine model makes it almost visually indistinguishable from a real vertebral column. The flexible vertebral column including occipital plate, pelvis and sacroccyx is mounted on a stand. Within the spinal canal, the spinal cord with caudal equina and exiting nerve roots are represented in flexible material.
Accessories information: For information about 3B smart anatomy, please see the document attached on this page.