ContactUsLinkComponent
Franck-Hertz Experiment Power Supply
Franck-Hertz Experiment Power Supply
Catalog # 470137-702
undefined
Franck-Hertz Experiment Power Supply
Catalog # 470137-702
Supplier Number:  1012818
Specifications
  • Description:
    Franck-Hertz Experiment Power Supply
  • Cat. no.:
    470137-702
  • Supplier No.:
    U8482530-115
Specifications
About this item
Adjustable Voltage Power Supply
This is the power supply unit for operating the mercury-filled Franck-Hertz tube and the neon-filled Franck-Hertz tube. It provides all the voltages needed to power the tubes and includes a sensitive built-in DC amplifier for measuring collector current. The accelerating voltage can be set-up manually on the apparatus or set to a saw-tooth wave form. Additional measuring inputs are also available for the anode current and accelerating voltage.