Air Sampling Filter Screens, Ace Glass Incorporated
Supplier: Ace Glass
Certificates
About this item
Filter screen made up of PTFE with 6.4 mm mesh is designed for use on the funnel end of air sampling canes.
The screen is held on to the bottom of the funnel by a stainless steel yoke retainer and three springs. Screen size is 152 mm.
Ordering information: Available complete with screen and retainer, or those component items may be purchased separately.
Customers Who Bought This Also Bought