Heating Tops, Ace Glass
Supplier: Ace Glass
Certificates
About this item
Designed for use with 1-, 2-, and 3-neck flasks, these convenient heating tops can be placed onto a flask without disturbing permanently attached equipment
Heating tops offer a temperature range of ambient+10 to 450 °C (842 °F).
Certifications: All 115 V units (only) are CSA certified
Ordering information: Heating tops are designed for use with heating mantles, and must be operated through a temperature controller. 230 V versions of each heating top are available through special order. Contact your Avantor sales representative for details. Supplied with a detachable 122 cm (4'), two-wire cord, and locking connector.
Customers Who Bought This Also Bought