(±)-3-Methyl-3-heptanol 98%
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:5582-82-1
:
Formula:C₈H₁₈O
MDL Number:MFCD00021841
Boiling Pt:64 °C/201 mm
Density:0.83
Molecular Weight:130.23 g/mol
Flash Pt:58 °C (136 °F)
Storage Temperature:Ambient
UN:1987
ADR:3 III
:GHS02
:GHS07
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Related Information
Certificates
Search by Lot NumberAlfa Aesar_Buy 3 Get 2 Complimentary items
Promotion Material
Organic Building Blocks and Intermediates
Application Notes
Frequently Bought Together