Indium, AAS Standard Solution, Specpure®, In 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
Certificates
About this item
Molecular Formula: In in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134048
Related Information
Certificates
Search by Lot NumberAlfa Aesar_Buy 3 Get 2 Complimentary items
Promotion Material
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together