Ammonium, Ion chromatography standard solution, Specpure®, NH4+ 1000µg/ml, Thermo Scientific
Catalog # CAAA35560-AE
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Concentration:1000 ppm
- Description:Ammonium standard solution
- Size:100 ml
- Cat. no.:CAAA35560-AE
- Contents:(NH₄)₂SO₄ in H₂O
- Supplier No.:35560-AE
Specifications
About this item
Standard from (NH₄)₂SO₄ in H₂O
Traceable to NIST