Order Entry
Canada
ContactUsLinkComponent
VWR® RiboReady™ Color RNA Ladders
VWR® RiboReady™ Color RNA Ladders
Catalog # 89239-528
Supplier:  VWR
UL Listed
CAS Number:  
undefined
VWR® RiboReady™ Color RNA Ladders
Catalog # 89239-528
Supplier:  VWR
Supplier Number:  1B1492-25RXN
UL Listed
CAS Number:  

Specifications

  • Description:
    RiboReady™ Color 1KB RNA Ladder
  • Cat. no.:
    89239-528
  • Application:
    25 lanes
  • Supplier No.:
    1B1492-25RXN

Specifications

About this item

RiboReady™ Color Micro RNA Ladder is made for precise sizing of RNA molecules in denaturing acrylamide gels

RiboReady™ Color 1KB RNA Ladder is made for precise sizing of RNA molecules in denaturing agarose gels. RiboReady™ Color 1KB RNA Ladder is supplied in a ready-to-use mixture that does not require heating or addition of a denaturing agent before use. RiboReady™ Color 1KB RNA Ladder has the added advantage of containing pre-stained single-strand nucleic acids that are visible during electrophoresis. The apparent sizes of bands in the RiboReady™ Color 1KB RNA Ladder are 200, 500, 1,000, 2,000, 4,000 and 8,000 bases in length.

RiboReady™ Color Micro RNA Ladder is supplied in a ready-to-use mixture that does not require heating or addition of a denaturing agent before use. RiboReady™ Color Micro RNA Ladder has the added advantage of containing pre-stained single-strand nucleic acids that are visible during electrophoresis. The apparent sizes of bands in the RiboReady™ Color Micro RNA Ladder are 100, 75, 50, 40, 30 and 20 bases in length.