ContactUsLinkComponent
Receiver, Moisture Test, Bidwell and Sterling-Type, Ace Glass
Receiver, Moisture Test, Bidwell and Sterling-Type, Ace Glass
Catalog # 89060-336
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Receiver, Moisture Test, Bidwell and Sterling-Type, Ace Glass
Catalog # 89060-336
Supplier:  Ace Glass
Supplier Number:  7705-02
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • Capacity:
    5 ml
  • Cat. no.:
    89060-336
  • ST Joint Size:
    24/40
  • Supplier No.:
    7705-02
Specifications
About this item
This Bidwell and Sterling style moisture test receiver is used to determine the moisture content in foods, fatty acids, etc
Fabricated from borosilicate glass.
Certifications: Conforms to specifications of ASTM and A.O.A.C.